Spécifications
- Description:Simulated Blood Transfusion Refill Kit
- No. of Lab Groups:6
- Cat. no.:470023-688
- Supplier No.:WARD470023-688
Spécifications
A propos de cet article
In complete safety, this kit teaches students how to determine blood types.
- Pre- and Post-Laboratory Assessments Included
- Kit Introduces Tests Essential to Successful Blood Transfusions
- Tests Link to Real-World Concepts and Professional Practices
- Two-Part Lab, 30 Minutes Each
- Material for Six Setups
They perform the same tests hematologists use. Students also practice cross-matching, which is vital for successful blood transfusions.
Students use Ward's Simulated Blood to re-create ABO blood-typing techniques practiced in real labs and determine what types are compatible for transfusions. This kit includes a teacher’s guide, a student copymaster, and enough material for six setups.