ContactUsLinkComponent
WARDS SIM. BLOOD TRANSFUSION LAB REFILL
Ward's® Simulated Blood Transfusion Matching Kit
# de catalogue 470023-688
Fournisseur:  Avantor
WARDS SIM. BLOOD TRANSFUSION LAB REFILL
Ward's® Simulated Blood Transfusion Matching Kit
# de catalogue 470023-688
Fournisseur:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Simulated Blood Transfusion Refill Kit
  • No. of Lab Groups:
    6
  • Cat. no.:
    470023-688
  • Supplier No.:
    WARD470023-688
Spécifications
A propos de cet article
In complete safety, this kit teaches students how to determine blood types.
  • Pre- and Post-Laboratory Assessments Included
  • Kit Introduces Tests Essential to Successful Blood Transfusions
  • Tests Link to Real-World Concepts and Professional Practices
  • Two-Part Lab, 30 Minutes Each
  • Material for Six Setups
They perform the same tests hematologists use. Students also practice cross-matching, which is vital for successful blood transfusions.
Students use Ward's Simulated Blood to re-create ABO blood-typing techniques practiced in real labs and determine what types are compatible for transfusions. This kit includes a teacher’s guide, a student copymaster, and enough material for six setups.
Ordering information: This kit includes Simulated Blood of Recipient: Type B; Simulated Blood of Mr. Smith: Type A; Simulated Blood of Mr. Jones: Type B; Simulated Blood of Mr. Green: Type AB; Simulated Blood of Ms. Brown: Type O, simulated anti-A typing serum, simulated anti-A typing serum, simulated recipient serum, blood-typing slides, a package of toothpicks, and micro-centrifuge tubes. A micro-centrifuge is required but not included.
Customers Who Bought This Also Bought