Vermiculite
# de catalogue 470021-148
Fournisseur: Avantor
Spécifications
- Description:Vermiculite, 4 qts.
- Cat. no.:470021-148
- Supplier No.:WARD470021-148
Spécifications
A propos de cet article
Light, porous mica medium retains moisture well, making it excellent for starting seeds or as a soil additive.
Customers Who Bought This Also Bought