Ward's® Microhematocrit Set
# de catalogue 470019-794
Fournisseur: Avantor
Spécifications
- Description:WARD’S Microhematocrit Set
- Cat. no.:470019-794
- Supplier No.:WARD470019-794
Spécifications
A propos de cet article
The set includes 200 heparinized, mylar wrapped, hematocrit tubes, and six capillary tube trays with sealant. Each tray has two rows of slots numbered 1 through 24 for easy tube storage.
Customers Who Bought This Also Bought