Muscle (Mammalian), Striated Slide
# de catalogue 470007-802
Fournisseur: Avantor
Spécifications
- Description:Muscle (Mammalian), Striated
- Cat. no.:470007-802
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470007-802
- Orientation:Cross section & longitudinal section
Spécifications
A propos de cet article
Voluntary Muscle Type
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Entire small muscle shown.
Customers Who Bought This Also Bought