ContactUsLinkComponent
LAND SNAIL (HELIX) JAR/10 PRESERVED
Preserved Land Snail
# de catalogue 470001-326
Fournisseur:  Avantor
LAND SNAIL (HELIX) JAR/10 PRESERVED
Preserved Land Snail
# de catalogue 470001-326
Fournisseur:  Avantor
Spécifications
  • Description:
    Preserved Land Snail
  • Size:
    1"
  • Injection Type:
    Uninjected
  • Cat. no.:
    470001-326
  • Packaging:
    Jar of 10
  • Supplier No.:
    WARD470001-326
Spécifications
A propos de cet article
Formalin preserved land snails. Available in jars or pails.
  • Gastropod
  • Invertebrate anatomy
Helix sp. The body and eyestalks are fully extended. Shell size: approximately 1".
Customers Who Bought This Also Bought