Large Southern Centipede
# de catalogue 470001-278
Fournisseur: Avantor
Spécifications
- Description:Large Southern Centipede, Preserved, Jar of 10
- Cat. no.:470001-278
- Supplier No.:WARD470001-278
Spécifications
A propos de cet article
Alcohol Preserved
Scolopendra. This species has one pair of legs per body segment and large “poison claws”. Size: 3–5"L.
Customers Who Bought This Also Bought