Preserved Earthworms
# de catalogue 470001-236
Fournisseur: Avantor
Spécifications
- Size:9"+
- Injection Type:Uninjected
- Cat. no.:470001-236
- Packaging:Jar of 10
- Supplier No.:WARD470001-236
Spécifications
A propos de cet article
Formalin preserved earthworms. Available in vacuum packs or jars.
- Introduction to dissection
- Annelid anatomy
Earthworms, Lumbricus sp., are great for teaching dissection. Our earthworms are available in medium and large sizes. Plain preserved and single injected earthworms are available.
Customers Who Bought This Also Bought