ContactUsLinkComponent
GONIONEMUS PRESERVED JAR/10
Gonionemus
# de catalogue 470001-204
Fournisseur:  Avantor
GONIONEMUS PRESERVED JAR/10
Gonionemus
# de catalogue 470001-204
Fournisseur:  Avantor
Spécifications
  • Description:
    Gonionemus, Preserved, Jar of 10
  • Cat. no.:
    470001-204
  • Supplier No.:
    WARD470001-204
Spécifications
A propos de cet article
Formalin Preserved
This small cosmopolitan jellyfish clearly shows the structure of hydrozoan medusae. Size: 3/8".
Customers Who Bought This Also Bought