Order Entry
ContactUsLinkComponent
GRADUATED CYLINDER 500ML PP WIDE MOUTH
Wide-Mouth Polypropylene Cylinders
# de catalogue 470020-850
Fournisseur:  SI Manufacturing
GRADUATED CYLINDER 500ML PP WIDE MOUTH
Wide-Mouth Polypropylene Cylinders
# de catalogue 470020-850
Fournisseur:  SI Manufacturing

Spécifications

  • Capacity:
    500 ml
  • Form:
    Tall
  • Cat. no.:
    470020-850
  • Supplier No.:
    78125-A

Spécifications

A propos de cet article

Chemical and Heat Resistant to 80°C.

  • Wide mouth is ideal for performing density tests
  • Molded, blue over-printed graduations
  • Aligned with GLOBE protocols

The squat design increases stability on the bench while the low form requires less overhead storage space.

Customers Who Bought This Also Bought