Cotton-Tipped Swab Applicators, SciEd Rochester Catalog Divisi
# de catalogue 470019-172
Fournisseur: Avantor
Spécifications
- Length:6"
- Sterility:Sterile
- Cat. no.:470019-172
- Supplier No.:WARD470019-172
Spécifications
A propos de cet article
Variety of sterile or non sterile swabs.
Sterile swabs have wooden shafts and are indivdually sealed. Depending on the pack size ordered, they may be packaged in a bag (pack/10) or box (pack/100). Excellent for spreading a confluent lawn of bacteria without breaking the agar surface.
Non sterile swabs are packaged together in a box or bag. Excellent for manipulating small samples that do not need to be kept sterile.
Customers Who Bought This Also Bought