Liquid Convection Apparatus
# de catalogue 470005-410
Fournisseur: Avantor
Spécifications
- Description:Liquid Convection Apparatus
- Cat. no.:470005-410
- Supplier No.:WARD470005-410
Spécifications
A propos de cet article
The movement of liquid currents due to differences in temperature is demonstrated by introducing dye into the water in the tube. One corner of the tube is heated and the movement of the dye clearly establishes the currents. The rectangular shaped tube measures 15×21 cm and has an opening at the top.
Customers Who Bought This Also Bought