ContactUsLinkComponent
MARBLE RAMP TD/CT
Marble Ramp
# de catalogue 470003-992
Fournisseur:  Avantor
MARBLE RAMP TD/CT
Marble Ramp
# de catalogue 470003-992
Fournisseur:  Avantor
Spécifications
  • Description:
    Marble Ramp
  • Cat. no.:
    470003-992
  • Supplier No.:
    WARD470003-992
Spécifications
A propos de cet article
Ideal for students conduct experiments by varying the angle of the marble track as well as the height from which the marble is released.
  • Includes instructions
  • Grades 7 - 9
  • Size: 96.5 x 12.7cm
By gathering data and plotting a graph, students discover that the same result can often be achieved by different combinations of angle and height.
Caution: CHOKING HAZARD - Item contains a marble. Not for children under 3 yrs.
Customers Who Bought This Also Bought