Polystyrene Spot Plate
# de catalogue 470002-082
Fournisseur: Avantor
Spécifications
- Description:Polystyrene spot plate
- Supplier No.:WARD470002-082
Spécifications
A propos de cet article
Chemically inert and unbreakable.
- Can be used with concentrated acids
Each plate has 12 ¹/₃" diameter depressions. Size: 4¹/₂×3¹/₂"
Packaging: Package of 12.
Customers Who Bought This Also Bought