ContactUsLinkComponent
POLYSTYRENE COLOR PLATE PKG/12
Polystyrene Spot Plate
# de catalogue 470002-082
Fournisseur:  Avantor
POLYSTYRENE COLOR PLATE PKG/12
Polystyrene Spot Plate
# de catalogue 470002-082
Fournisseur:  Avantor
Spécifications
  • Description:
    Polystyrene spot plate
  • Supplier No.:
    WARD470002-082
Spécifications
A propos de cet article
Chemically inert and unbreakable.
  • Can be used with concentrated acids
Each plate has 12 ¹/₃" diameter depressions. Size: 4¹/₂×3¹/₂"
Packaging: Package of 12.
Customers Who Bought This Also Bought