ContactUsLinkComponent
PIG HEART PRESERVED VACPACK/1
Preserved Pig Hearts
# de catalogue 470001-654
Fournisseur:  Avantor
PIG HEART PRESERVED VACPACK/1
Preserved Pig Hearts
# de catalogue 470001-654
Fournisseur:  Avantor
Spécifications
  • Description:
    Preserved Pig Heart without Pericardium
  • Cat. no.:
    470001-654
  • Packaging:
    Vacuum pack of 1
  • Supplier No.:
    WARD470001-654
Spécifications
A propos de cet article
Formalin preserved pig hearts. Available in vacuum packs or pails. With or without pericardium. Shipped in Wardsafe
  • Mammalian organ dissection
  • With or without pericardium
  • Heart dissection
Our pig hearts are great teaching specimens to learn about the mammalian heart and cardiac anatomy. Pig hearts are larger than sheep hearts. Available with or without pericardium.
Customers Who Bought This Also Bought