ContactUsLinkComponent
PIG BRAIN WITH EYES PAIL/1
Pig Brain with Eyes
Catalog # 470000-920
Supplier:  Avantor
undefined
Pig Brain with Eyes
Catalog # 470000-920
Supplier:  Avantor
Specifications
  • Cat. no.:
    470000-920
  • Packaging:
    Jar of 1
  • Supplier No.:
    WARD470000-920
Specifications
About this item
Formalin preserved pig brain with eyes. Available in jars.
  • Mammalian anatomy
  • High quality preservation
Featuring intact eyes, dura mater, hypophysis, and some intact cranial nerves, this brain (which may lack the cerebellum) is ideal for demonstrations because it is slightly larger than the sheep brain.
Frequently Bought Together