Preserved Common Jellyfish
# de catalogue 470000-872
Fournisseur: Avantor
Spécifications
- Size:4" to 6"
- Cat. no.:470000-872
- Packaging:Pail
- Supplier No.:WARD470000-872
Spécifications
A propos de cet article
Formalin preserved common jellyfish. Packed in jars or pails.
- Jellyfish anatomy
- Invertebrate dissection
These common jellyfish (Aurelia) are an example of a scyphozoan.
Ordering information: Jellyfish are seasonal and typically available during the summer months, outside of the season, supply is limited.
Customers Who Bought This Also Bought