ContactUsLinkComponent
GREEN SEA URCHIN PL/10 PM
Green Sea Urchin
# de catalogue 470000-772
Fournisseur:  Avantor
GREEN SEA URCHIN PL/10 PM
Green Sea Urchin
# de catalogue 470000-772
Fournisseur:  Avantor
Spécifications
  • Description:
    Green Sea Urchin, Preserved, Pail of 10
  • Cat. no.:
    470000-772
  • Supplier No.:
    WARD470000-772
Spécifications
A propos de cet article
Formalin Preserved
Strongylocentrotus drobachiensis.Body size: 1–2".
Characterized by complex feeding structures with five calcareous protractable teeth (Aristotle’s lantern).
Customers Who Bought This Also Bought