Preserved Land Snail
# de catalogue 470000-754
Fournisseur: Avantor
Spécifications
- Description:Preserved Land Snail
- Size:1"
- Injection Type:Uninjected
- Cat. no.:470000-754
- Packaging:Pail of 100
- Supplier No.:WARD470000-754
Spécifications
A propos de cet article
Formalin preserved land snails. Available in jars or pails.
- Gastropod
- Invertebrate anatomy
Helix sp. The body and eyestalks are fully extended. Shell size: approximately 1".
Customers Who Bought This Also Bought