Pregnant Rats
# de catalogue 470000-586
Fournisseur: Ward's Science
Spécifications
- Description:Pregnant Rats
- Size:7 to 9”
- Injection Type:Double injected
- Cat. no.:470000-586
- Packaging:Pail of 10
- Supplier No.:WARD470000-586
Spécifications
A propos de cet article
Formalin preserved rats. Available in vacuum packs or pails.
- Mammalian anatomy
- General dissection
- Reproduction
Our rat specimens are carefully preserved in an extended position and are mature subjects for students to examine fully developed animals, their organ systems, and their skeletal, muscular, and reproductive structures. Pregnant rats are available as unijected or double injected. Injected rats are made without incisions in the neck, ensuring the specimens are intact. Sizes 7 to 9”.
Customers Who Bought This Also Bought