Preserved Snakes
470000-584
: Avantor
- Description:Preserved snakes
- Size:18"+
- Injection Type:Uninjected
- Cat. no.:470000-584
- Packaging:Pail of 10
- Supplier No.:WARD470000-584
Formalin preserved snake available in vacuum packs and pails.
- Repitile anatomy
- High quality preservation
Snakes are common reptiles used in dissection and to teach, among other things, vertebrate anatomy and comparative physiology. Students can explore the many vertebrae along their body in comparison to humans, who have just 32 vertebrae. Many snakes are camouflaged by color and pattern to blend into their surroundings and mimic their natural habitat. Garter snakes or other common non-poisonous species are supplied.
Customers Who Bought This Also Bought