Ward's® Preserved Bullfrogs: Double Injected
# de catalogue 470000-562
Fournisseur: Ward's Science
Spécifications
- Size:6 to 7"
- Injection Type:Double injected
- Cat. no.:470000-562
- Packaging:Pail of 10
- Supplier No.:WARD470000-562
Spécifications
A propos de cet article
Formalin preserved bullfrogs Available in vacuum packs or pails.
- Amphibian anatomy
- General dissection
- High quality preservation
Bullfrogs (Rana catesbeiana) offer a larger study specimen for students to learn vertebrate anatomy and are particularly good for younger students to practice their developing dissection skills. Their internal organs and structures are easy to see and are easily distinguishable.
Caution: Amphibian anatomy
General dissection
High quality preservation
Customers Who Bought This Also Bought