ContactUsLinkComponent
SINGLE INJECTED RAT PAIL/25 PRESERVED
Ward's® Preserved Rats
# de catalogue 470000-506
Fournisseur:  Ward's Science
Ward's® Preserved Rats
# de catalogue 470000-506
Fournisseur:  Ward's Science
Spécifications
  • Size:
    7 to 9”
  • Injection Type:
    Single injected
  • Cat. no.:
    470000-506
  • Packaging:
    Pail of 25
  • Supplier No.:
    WARD470000-506
Spécifications
A propos de cet article
Formalin preserved rats. Available in vacuum packs or pails.
  • Mammalian anatomy
  • General dissection
  • High quality preservation
Our rat specimens are carefully preserved in an extended position and are mature subjects for students to examine fully developed animals, their organ systems, and their skeletal, muscular, and reproductive structures. We carry plain preserved, single, double, and triple injected rats. Injected rats are made without incisions in the neck, ensuring the specimens are intact. Sizes 7 to 9”.
Customers Who Bought This Also Bought