Ward's® Live Euglena sp. (gracilis)
Catalog # 470189-298
Supplier: Boreal Laboratories
Specifications
- Description:Euglena sp. Study Set
- Cat. no.:470189-298
- Media:Euglena medium
- No. of Students:30
- Storage Conditions:Room temp, high light, loose cap
- Packaging:Jar
- Supplier No.:WARD470189-298
Specifications
About this item
Characterized by a lack of rigid cellulosic walls, instead these organisms have pellicles made of protein. They store paramylon instead of starch.
- Excellent for studying general flagellates
Study Set contains: a culture of Euglena sp. And a Form and Structure Copymaster diagram
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.
Customers Who Bought This Also Bought