Live Amoeba proteus cultures
# de catalogue 470189-294
Fournisseur: Boreal Laboratories
Spécifications
- Description:Amoeba proteus Study Set for 30 Students
- Storage Conditions:Room temp, loose cap
- No. of Students:30
- Cat. no.:470189-294
- Packaging:comes bundled with our form and structure sheet, and a pipet to transfer the amoeba
- Media:Amoeba medium
- Supplier No.:WARD470189-294
Spécifications
A propos de cet article
Demonstrate amoeba structure, locomotion, and feeding behavior.
- Demonstrate amoeba structure, locomotion, and feeding behavior
- ReadySlides are prepared for you you and offer a convenient, ready-to-go option
- Vital Stained cultures allow students to better visualize organism structures
Amoeba proteus is a great sarcodine for demonstrating amoebae structure. They use pseudopods for both locamotion and feeding. Amoeba feed on Chilomonas, or other small protozoa, which they phagocytize with their pseudopodia. Amoeba cultures are supplied in Amoeba Media with a rice grain and food (Chilomonas).
Ordering information: These products contain living or perishable materials. Please specify a delivery date.
Customers Who Bought This Also Bought