Wire Leads with Alligator Clips
# de catalogue 470189-160
Fournisseur: United Scientific Supplies
Spécifications
- Description:Wire Leads, Black, w/ Alligator Clips
- Cat. no.:470189-160
- Supplier No.:WAG024 BLK
Spécifications
A propos de cet article
Easily connect electrical devices that are lacking banana plugs.
These wire leads are extra flexible and are plastic coated with alligator clips at both ends. The leads are 56 cm long and the alligator clips feature a protective vinyl sheath.
- Flexible plastic coated wire
- 18 gauge wire
These wire leads are extra flexible and are plastic coated with alligator clips at both ends. The leads are 56 cm long and the alligator clips feature a protective vinyl sheath.
Customers Who Bought This Also Bought