Rubber Policeman
# de catalogue 470189-046
Fournisseur: United Scientific Supplies
Spécifications
- Description:Without glass rod
- Cat. no.:470189-046
- Supplier No.:RHGSR1
Spécifications
A propos de cet article
Ideal for Scraping and Mixing.
- Use as a policeman, stirring rod, or as a spatula
Rubber policeman is 15 mm natural rubber, fits 5 and 6 mm Ø rods. Glass rod is 15 cm long.
Customers Who Bought This Also Bought