Cartilage, Composite Slide
# de catalogue 470182-642
Fournisseur: Avantor
Spécifications
- Description:Cartilage, Composite Slide
- Cat. no.:470182-642
- Stain:Verhoeff and eosin
- Supplier No.:WARD470182-642
- Orientation:Section
Spécifications
A propos de cet article
Investigate the characteristics of cartilage with and without elastic fibers
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Section of hyaline and elastic cartilage showing the presence and absence of elastic fibers.
Customers Who Bought This Also Bought