Pseudostratified, Ciliated, Columnar Epithelium Slides
# de catalogue 470182-616
Fournisseur: Avantor
Spécifications
- Description:Pseudostrat, Cil. Col. Epithelium
- Cat. no.:470182-616
- Stain:Masson
- Supplier No.:WARD470182-616
- Orientation:Section
Spécifications
A propos de cet article
Demonstrates tissues in the upper respiratory tract that helps trap foreign bodies.
- Identifying features clearly distinguishable
- Rigorous quality control standards
Choose from several available stains for these tissue sections to get the view you're looking for.
Customers Who Bought This Also Bought