Toxocara canis, Eggs Slide
Catalog # 470182-184
Supplier: Avantor
Specifications
- Description:Toxocara canis, Eggs
- Cat. no.:470182-184
- Stain:Giemsa
- Supplier No.:WARD470182-184
- Orientation:Whole mount
Specifications
About this item
A Common Parasite in Canids
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ingestion of these embryonated eggs is responsible for visceral larva migrans in humans. Unstained. The Toxocara canis slide is double cover mounted, and therefore is best viewed using 4 and 10x microscope objectives.
Customers Who Bought This Also Bought