Metridium Composite Slide
# de catalogue 470181-544
Fournisseur: Avantor
Spécifications
- Description:Metridium Composite
- Cat. no.:470181-544
- Stain:Mallory triple
- Supplier No.:WARD470181-544
- Orientation:Cross section & longitudinal section
Spécifications
A propos de cet article
Plumose Sea Anemone
- Identifying features clearly distinguishable
- Rigorous quality control standards
Comparative study of diploblastic body plan.
Customers Who Bought This Also Bought