ContactUsLinkComponent
METRIDIUM-COMPOSITE (CS/LS) MAL
Metridium Composite Slide
# de catalogue 470181-544
Fournisseur:  Avantor
METRIDIUM-COMPOSITE (CS/LS) MAL
Metridium Composite Slide
# de catalogue 470181-544
Fournisseur:  Avantor
Spécifications
  • Description:
    Metridium Composite
  • Cat. no.:
    470181-544
  • Stain:
    Mallory triple
  • Supplier No.:
    WARD470181-544
  • Orientation:
    Cross section & longitudinal section
Spécifications
A propos de cet article
Plumose Sea Anemone
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Comparative study of diploblastic body plan.
Customers Who Bought This Also Bought