ContactUsLinkComponent
AURELIA-SCYPHISTOMA (WM)
Aurelia, Scyphistoma Slide
# de catalogue 470181-534
Fournisseur:  Avantor
AURELIA-SCYPHISTOMA (WM)
Aurelia, Scyphistoma Slide
# de catalogue 470181-534
Fournisseur:  Avantor
Spécifications
  • Description:
    Aurelia, Scyphistoma
  • Cat. no.:
    470181-534
  • Stain:
    Schneider's and fast green
  • Supplier No.:
    WARD470181-534
  • Orientation:
    Whole mount
Spécifications
A propos de cet article
Polyp Form
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Asexual stage that undergoes strobilization, or asexual reproduction by transverse fission, to give rise to several ephyrula.
Customers Who Bought This Also Bought