Aurelia, Scyphistoma Slide
# de catalogue 470181-534
Fournisseur: Avantor
Spécifications
- Description:Aurelia, Scyphistoma
- Cat. no.:470181-534
- Stain:Schneider's and fast green
- Supplier No.:WARD470181-534
- Orientation:Whole mount
Spécifications
A propos de cet article
Polyp Form
- Identifying features clearly distinguishable
- Rigorous quality control standards
Asexual stage that undergoes strobilization, or asexual reproduction by transverse fission, to give rise to several ephyrula.
Customers Who Bought This Also Bought