Euglena Slide
# de catalogue 470181-458
Fournisseur: Avantor
Spécifications
- Description:Euglena
- Cat. no.:470181-458
- Stain:Fast Green and Mallory's
- Orientation:Whole Mount
Spécifications
A propos de cet article
Single Celled Flagellates
- Identifying features clearly distinguishable
- Rigorous quality control standards
Large euglenoid which shows discoid chromatophores, paramylon bodies, and stigma.
Customers Who Bought This Also Bought