ContactUsLinkComponent
PLATESPOTATO-DEXTROSE AGAR PKG/10
Ward's® Prepared Potato Dextrose Agar
Catalog # 470180-690
CAS Number:  
PLATESPOTATO-DEXTROSE AGAR PKG/10
Ward's® Prepared Potato Dextrose Agar
Catalog # 470180-690
CAS Number:  
Specifications
  • Pack type:
    10 plates
  • Description:
    Agar plates
  • Cat. no.:
    470180-690
  • Supplier No.:
    880915
Specifications
About this item
For growing bacteria and fungi
  • Sterile- no autoclaving
  • Convenient - prepared for you
  • Prepared consistenly every time
Potato dextrose agar (or PDA agar) is a microbiological growth media made from potato infusion and dextrose.
Ordering information: Some of these products contain living or perishable materials
Delivery: Prepared plates will ship via overnight or 2nd day delivery.
Prepared bottles and tubes do not require as much temperature regulation in-transit so we ship them via ground to save you the cost of freight. During checkout, please specify a delivery date for your specimens to arrive.
Customers Who Bought This Also Bought