Ward's® Live Green Tree Frog (Hyla sp.)
# de catalogue 470180-580
Fournisseur: STRICTLY REPTILES
Spécifications
- Description:Green Tree Frog
- Cat. no.:470180-580
- Body Size:1.0 - 1.5"
- Supplier No.:WARD470180-580
Spécifications
A propos de cet article
These fascinating frogs are uniquely adapted for climbing, with prominent adhesive pads on the ends of their digits.
- Makes for a great lab or classroom pet.
- Will occasionally vocalize, especially during springtime.
Keep in glass or plastic terrarium. Will feed on small insects such as cricket nymphs or flightless fruit flies.
Ordering Information: Care instructions for these frogs are available online.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought