ContactUsLinkComponent
DROSOPHILA MOJAVENSIS-LIVE CULTURE
Drosophila mojavensis
# de catalogue 470180-452
Fournisseur:  Ward's Science
DROSOPHILA MOJAVENSIS-LIVE CULTURE
Drosophila mojavensis
# de catalogue 470180-452
Fournisseur:  Ward's Science
Spécifications
  • Description:
    Drosophila mojavensis, For 35 Students
  • Cat. no.:
    470180-452
  • Supplier No.:
    WARD470180-452
Spécifications
A propos de cet article
Long life cycle (approximately one month). Useful for comparative population studies and for preparing salivary gland chromosome squashes.
Recommended by the Carnegie Institute
Mature Drosophila are cultured in our own labs, and are marked with the culture initiation date. Culture is shipped in our instant medium in a shatterproof vial. The pupae will usually appear within ten days of the ship date. You will also receive WARD’S Drosophila Culture Guide. Note: We can supply any of our single strains in the larger quantity for 100 students. Call our Customer Service department for details.
Customers Who Bought This Also Bought