Ward's® Drosophila Culturing Set
# de catalogue 470180-400
Fournisseur: Avantor
Spécifications
- Description:WARD’S Drosophila Culturing Set
- Cat. no.:470180-400
- Supplier No.:WARD470180-400
Spécifications
A propos de cet article
Readily Observed Strain Requires No Magnification
Use this unique culturing set as a convenient introduction to genetics; students can perform a simple monohybrid cross to yield a 3:1 winged: wingless ratio. Females have red eyes with yellow bodies, males have white eyes with gray bodies, making them easy to distinguish. Cultures supplied are yellow-forked-attached and white, and yellow-forked-attached and white apterous. The set comes with WARD’S Drosophila Culture Guide and cultures for up to 15 setups.
Customers Who Bought This Also Bought