Ward's® Live Silkworm Eggs (Bombyx mori)
Catalog # 470180-382
Supplier: COASTAL SILKWORMS
Specifications
- Description:Artificial Silkworm Diet, 1/2 lb.
- Cat. no.:470180-382
- Supplier No.:WARD470180-382
Specifications
About this item
When hatched, the small, gray larvae are the traditional source for high-quality silk.
- They can remain refrigerated for up to a year, allowing for easy storage and access
- Eggs will hatch in 10-14 days if kept at 75-85°F
Over thousands of years of domestication and selective breeding the modern silkworm has lost its ability to fly. Eggs, each roughly the size of a pinhead, come glued onto a petri dish.
Ordering information: These products contain living or perishable materials.
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Frequently Bought Together