Ward's® Live Water Fleas (Daphnia magna)
# de catalogue 470180-304
Fournisseur: Avantor
Spécifications
- Description:Daphnia study set
- No. of Students:30
- Cat. no.:470180-304
- Supplier No.:WARD470180-304
Spécifications
A propos de cet article
Daphnia are a widely studied small crustacean used for a number of physiology studies including the effects of drugs on heartbeat and temperature on metabolism.
- Excellent source of food for small invertebrates amphibians
- Easy and safe to handle, great for observation
Daphnia magna belong to the order Cladocera. Can be kept for short periods of time at room temperature, with the cap loosened. Use only dechlorinated tap water or spring water.
Ordering information: These products contain living or perishable materials. Please specify a delivery date
Customers Who Bought This Also Bought