Ward's® Live Adult Brine Shrimp (Artemia sp.)
# de catalogue 470180-300
Fournisseur: SEGREST FARMS
Spécifications
- Description:Adult Brine Shrimp
- Storage Conditions:Room temp, loose cap
- No. of Students:30
- Cat. no.:470180-300
- Media:Salt water
- Supplier No.:WARD470180-300
Spécifications
A propos de cet article
These small marine crustaceans are supplied as adults, and measure approximately 1/4" in size at the time of shipment.
- Excellent food source for fish
- Great model for phototaxis
Ordering information: These products contain living or perishable materials.
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought