Water Bear
Catalog # 470180-260
Supplier: Ward's Science
Specifications
- Description:Water Bear, For 35 Students
- Cat. no.:470180-260
- Supplier No.:WARD470180-260
Specifications
About this item
Hypsibius sp. The water bear is a complex animal representing the unusual phylum of Tardigrades, which contains about 400 marine, terrestrial, and freshwater species.
- Unusual phylum
- Complex animals
- Can survive in adverse conditions
These complex animals can survive extremely adverse conditions, making them ideal research subjects for studies on the aging process. Culture instructions are included. Size: up to 1 mm.
Customers Who Bought This Also Bought