Soil Nematodes
# de catalogue 470180-246
Fournisseur: Ward's Science
Spécifications
- Description:Nematodes,Soil
- No. of Students:30
- Cat. no.:470180-246
- Packaging:Jar
- Media:Soil water medium
- Supplier No.:WARD470180-246
Spécifications
A propos de cet article
Examine the morphological characteristics of typical soil nematodes with the specimens in our well-populated culture.
- Keeps for up to two months if refrigerated
- Examines morphological characteristics
- Well-populated culture
Examine the morphological characteristics of typical soil nematodes with the specimens in our well-populated culture. Refrigerate the culture at 48°F and it should keep for up to two months. Culture instructions are included. Size: 0.5–5 mm.
Customers Who Bought This Also Bought