Live Paramecium aurelia
# de catalogue 470180-150
Fournisseur: Avantor
Spécifications
- Description:Paramecium aurelia Culture for 100 Students
- Cat. no.:470180-150
- Supplier No.:WARD470180-150
Spécifications
A propos de cet article
Paramecium aurelia is the smallest species of Paramecium we offer. Paramecium aurelia is characterized by cilia, which it uses for locomotion and feeing. Contains a macronucleus and two micronuclei. Grows in Dilute Alfalfa Medium (Hay Medium).
Customers Who Bought This Also Bought