ContactUsLinkComponent
NAVICULA PELLICULOSA 35 STUDENTS
Navicula
Catalog # 470179-846
Supplier:  Ward's Science
NAVICULA PELLICULOSA 35 STUDENTS
Navicula
Catalog # 470179-846
Supplier:  Ward's Science
Specifications
  • Description:
    Navicula pelliculosa (Small)
  • Cat. no.:
    470179-846
  • Media:
    Basic culture solution
  • No. of Students:
    30
  • Packaging:
    Tube
  • Supplier No.:
    WARD470179-846
Specifications
About this item
Freshwater. Unicellular; pennate, with bilateral symmetry. Basic culture solution.
Characterized by shells (tests) made of silica, in two parts.
Customers Who Bought This Also Bought