ContactUsLinkComponent
C. REINHARDTII (-) BROTH 100 STUDENTS
Ward's® Live Chlamydomonas reinhardtii Cultures
# de catalogue 470179-744
Fournisseur:  Avantor
C. REINHARDTII (-) BROTH 100 STUDENTS
Ward's® Live Chlamydomonas reinhardtii Cultures
# de catalogue 470179-744
Fournisseur:  Avantor
Spécifications
  • Description:
    C. reinhardtii (-) (Large)
  • Storage Conditions:
    Room temp, high light, loose cap
  • No. of Students:
    100
  • Cat. no.:
    470179-744
  • Packaging:
    Tube
  • Media:
    Basic culture solution
  • Supplier No.:
    WARD470179-744
Spécifications
A propos de cet article
Demonstrates isogamous reproduction where gametes are similar in appearance and behavior. Make lectures and labs on protist reproduction clearer with our specially selected cultures that illustrate various reproductive phenomona.
  • Both mating types (+) and (-) available
  • Bacteria-free (Axenic) cultures available for both mating types
This green alga is a unicellular flagellate commonly found in damp soil, and aquatic environments. Each cell has 2 flagella and a large eyespot that allow it to sense light.
While all genera of Chlamydomonas are capable of growing in the light, C. reinhartdii can also survive in the dark if it is provided organic carbon.
Genus: Chlamydomonas Species: reinhardtii
Customers Who Bought This Also Bought