ContactUsLinkComponent
CARTERIA 35 STUDENTS
Carteria
Catalog # 470179-720
Supplier:  Avantor
CARTERIA 35 STUDENTS
Carteria
Catalog # 470179-720
Supplier:  Avantor
Specifications
  • Description:
    Carteria (Small)
  • Cat. no.:
    470179-720
  • Media:
    Basic culture solution
  • No. of Students:
    30
  • Packaging:
    Tube
  • Supplier No.:
    WARD470179-720
Specifications
About this item
Freshwater. Unicellular; large cells, with four flagella. Basic culture solution.
Characterized by green plastids and cell walls composed of cellulose.
Customers Who Bought This Also Bought