To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Pseudomonas aeruginosa
- Cat. no.:470179-204
- Packaging:Tube
- Media:Tryptic soy agar
- Supplier No.:WARD470179-204
- Incubation Conditions:37 °C
Spécifications
A propos de cet article
This organism can be isolated from many human urinary tract infections, as well as polluted water and sewage. It is commonly identified by its distinctive grape-like smell (trimethylamine) or because it can turn the agar light green due to production of a blue pigment (pyocyanin).
- Biosafety Level: 2
- Canada: Containment Level 2 - PHAC Licence required
- Gram Stain: Negative
- Morphology: rods
- Motility: Positive
- Respiration: Aerobic
- Chromogenesis: Yes (blue)
All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage.
Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Bacterial and fungal cultures are restricted and can only be ordered by educational and research professionals.
Ward's Science is not affiliated with the American Type Culture Collection and does not provide corresponding ATCC numbers for our organisms.
As with all organisms, Ward's recommends that you utilize proper aseptic technique when handling all microorganisms.