Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Ward's® Live Enterococcus faecalis Culture (Pathogenic) | Avantor

Ward's® Live Enterococcus faecalis Culture (Pathogenic)

Enterococcus faecalis, Blood agar/tryptic soy agar, 37 °C, Tube
Catalog # 470179-184
Supplier:   Avantor
Ward's® Live <i>Enterococcus faecalis</i> Culture (Pathogenic)
Catalog # 470179-184
Supplier:  Avantor
Ward's® Live <i>Enterococcus faecalis</i> Culture (Pathogenic)
Ward's® Live Enterococcus faecalis Culture (Pathogenic)

Enterococcus faecalis, Blood agar/tryptic soy agar, 37 °C, Tube

Price
$22.05
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

This organism is normal flora of human intestine and may be pathogenic in humans. This organism will grow in Sodium chloride solution.

  • Biosafety Level: 2
  • Canada: Containment Level 2 - PHAC Licence required
  • Gram Stain: Positive
  • Morphology: cocci in short chains
  • Lancefiled Group: D
  • Bile esculin: Positive
  • Motility: Negative
  • Respiration: Aerobic
  • Hemolysis: Gamma

All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.

This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage.

Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.

Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.

Bacterial and fungal cultures are restricted and can only be ordered by educational and research professionals.

Ward's Science is not affiliated with the American Type Culture Collection and does not provide corresponding ATCC numbers for our organisms.

Caution: This organism is pathogenic. Ward's adheres to the Biosafety Guidelines for Microbiological and Biomedical Laboratories, as published by the National Institutes of Health.

As with all organisms, Ward's recommends that you utilize proper aseptic technique when handling all microorganisms.

Spécifications
  • Désignation:
    Enterococcus faecalis
  • Packaging:
    Tube
  • Media:
    Blood agar/tryptic soy agar
  • Incubation Conditions:
    37 °C

Certains produits peuvent apparaître comme restreints

Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.

Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)

 

Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.