ContactUsLinkComponent
LM ESCHERICHIA COLI K12 (G-ROD)
Ward's® Live E. coli cultures for Coliphage Use
# de catalogue 470179-082
Fournisseur:  Avantor
Ward's® Live E. coli cultures for Coliphage Use
# de catalogue 470179-082
Fournisseur:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Escherichia coli - Strain K12
  • Storage Conditions:
    Refrigerator
  • Cat. no.:
    470179-082
  • Packaging:
    Tube
  • Media:
    Tryptic soy agar
  • Supplier No.:
    WARD470179-082
  • Incubation Conditions:
    37 °C
Spécifications
A propos de cet article
A collection of some of the strains of Escherichia coli needed to grow various coliphages (available separately).
The successful culture of a large volume of coliphage requires growing the appropriate host bacterium in tryptic soy broth at 37 °C. Several strains of host bacteria are available.
Ordering information: Strain B: Optimum host for all T phages.
Strain C: Optimum host for PhiX coliphages; will also support T2 growth.
Strain K12: Lysogenic for coliphage lambda; supports reproduction of wild type T4, resulting in lysis. T4rII mutants do not form plaques. Host for T4rllA and T4rllB phages. Lysogenic for lambda phages. Used to study point mutations in cistron A and B.
Strain: W3104: Lysogenic for coliphage lambda. Ultraviolet radiation is effective in prophage induction.
Bacterial and fungal cultures are restricted and can only be ordered by educational professionals.
Caution: Although this organism is non pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.
Customers Who Bought This Also Bought