Ward's® Live E. coli cultures for Coliphage Use | Avantor
Ward's® Live E. coli cultures for Coliphage Use
Escherichia coli - Strain C, Tryptic soy agar, 37 °C, Refrigerator, Tube
Catalog # 470179-064
Supplier:   Ward's Science
Fournisseur #:  CALCOLIC
Ward's® Live<i> E. coli </i>cultures for Coliphage Use
Catalog # 470179-064
Supplier:  Ward's Science
Ward's® Live<i> E. coli </i>cultures for Coliphage Use
Ward's® Live E. coli cultures for Coliphage Use
Escherichia coli - Strain C, Tryptic soy agar, 37 °C, Refrigerator, Tube
Price
$22.05
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
A collection of some of the strains of Escherichia coli needed to grow various coliphages (available separately).
The successful culture of a large volume of coliphage requires growing the appropriate host bacterium in tryptic soy broth at 37 °C. Several strains of host bacteria are available.
Ordering information: Strain B: Optimum host for all T phages.
Strain C: Optimum host for PhiX coliphages; will also support T2 growth.
Strain K12: Lysogenic for coliphage lambda; supports reproduction of wild type T4, resulting in lysis. T4rII mutants do not form plaques. Host for T4rllA and T4rllB phages. Lysogenic for lambda phages. Used to study point mutations in cistron A and B.
Strain: W3104: Lysogenic for coliphage lambda. Ultraviolet radiation is effective in prophage induction.
Bacterial and fungal cultures are restricted and can only be ordered by educational professionals.
Caution: Although this organism is non pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.
Spécifications
  • Désignation:
    Escherichia coli - Strain C
  • Storage Conditions:
    Refrigerator
  • Packaging:
    Tube
  • Media:
    Tryptic soy agar
  • Incubation Conditions:
    37 °C