Iron-Deficiency Anemia Slide
# de catalogue 470177-660
Fournisseur: Avantor
Spécifications
- Description:Iron-Deficiency Anemia Slide
- Cat. no.:470177-660
- Stain:Giemsa
- Supplier No.:WARD470177-660
- Orientation:Smear
Spécifications
A propos de cet article
Illustrates red blood cell characteristics of the deficiency
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Peripheral blood. Characterized by small, pale red blood cells.
Customers Who Bought This Also Bought